Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyp2b1, Mouse, Monoclonal Antibody, Abnova™

Catalog No. 89111609 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-609 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-609 Supplier Abnova Corporation Supplier No. MAB3815
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Cyp2b1.

Specifications

Antigen Cyp2b1
Applications ELISA, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Cyp2b1.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In purified antibody
Gene Cyp2b1
Gene Alias Cyp2b-1/Cypbe/MGC188065
Gene Symbols Cyp2b1
Host Species Mouse
Immunogen Rat Cyp2b1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24300
Test Specificity Identification of 2B1 by immunoassay or Western blots, without cross-reactivity with 2B2 or 2A1.
Target Species Rat
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.