Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyp3a23/3a1, Mouse, Clone: 2.13.1, Abnova™

Catalog No. 89111610 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-610 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-610 Supplier Abnova Corporation Supplier No. MAB3818
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Cyp3a23/3a1.

Specifications

Antigen Cyp3a23/3a1
Applications Western Blot
Classification Monoclonal
Clone 2.13.1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Cyp3a23/3a1.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In purified antibody
Gene Cyp3a23/3a1
Gene Alias CYP/CYP3A23/Cyp3a1/Cyp3a3/MGC108757/RL33/cDEX
Gene Symbols Cyp3a23/3a1
Host Species Mouse
Immunogen Rat Cyp3a23/3a1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25642
Target Species Rat
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.