Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP4Z1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107690 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-690 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-690 Supplier Abnova Corporation Supplier No. H00199974B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human CYP4Z1 protein.

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This gene is part of a cluster of cytochrome P450 genes on chromosome 1p33. [provided by RefSeq

Sequence: MEPSWLQELMAHPFLLLILLCMSLLLFQVIRLYQRRRWMIRALHLFPAPPAHWFYGHKEFYPVKEFEVYHKLMEKYPCAVPLWVGPFTMFFSVHDPDYAKILLKRQDPKSAVSHKILESWVGRGLVTLDGSKWKKHRQIVKPGFNISILKIFITMMSESVRMMLNKWEEHIAQNSRLELFQHVSLMTLDSIMKCAFSHQGSIQLDSTLDSYLKAVFNLSKISNQRMNNFLHHNDLVFKFSSQGQIFSKFNQELHQFTEKVIQDRKESLKDKLKQDTTQKRRWDFLDILLSAKSENTKDFSEADLQAEVKTFMFAGHDTTSSAISWILYCLAKYPEHQQRCRDEIRELLGDGSSITWEHLSQMPYTTMCIKECLRLYAPVVNISRLLDKPITFPDGRSLPAGITVFINIWALHHNPYFWEDPQVFNPLRFSRENSEKIHPYAFIPFSAGLRNCIGQHFAIIECKVAVALTLLRFKLAPDHSRPPQPVRQVVLKSKNGIHVFAKKVC

Specifications

Antigen CYP4Z1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human CYP4Z1 protein.
Formulation PBS with no preservative; pH 7.4
Gene CYP4Z1
Gene Accession No. NM_178134.2
Gene Alias CYP4A20
Gene Symbols CYP4Z1
Host Species Mouse
Immunogen CYP4Z1 (NP_835235.1,1 a.a. ∽ 505 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 199974
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.