Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP7A1, Mouse, Clone: 8F1, Abnova™

Catalog No. 89104366 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-366 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-366 Supplier Abnova Corporation Supplier No. H00001581M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant CYP7A1.

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This endoplasmic reticulum membrane protein catalyzes the first reaction in the cholesterol catabolic pathway in the liver, which converts cholesterol to bile acids. This reaction is the rate limiting step and the major site of regulation of bile acid synthesis, which is the primary mechanism for the removal of cholesterol from the body. [provided by RefSeq

Sequence: MFEAGYLTIFGRDLTRRDTQKAHILNNLDNFKQFDKVFPALVAGLPIHMFRTAHNAREKLAESLRHENLQKRESISELISLRMFLNDTLSTFDDLEKAK

Specifications

Antigen CYP7A1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 8F1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant CYP7A1.
Formulation PBS with no preservative; pH 7.4
Gene CYP7A1
Gene Accession No. NM_000780.3
Gene Alias CP7A/CYP7/MGC126826/MGC138389
Gene Symbols CYP7A1
Host Species Mouse
Immunogen CYP7A1 (NP_000771.2,179 a.a. ∽ 277 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A Purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1581
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.