Learn More
CYTH3, Mouse, Clone: S1, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant CYTH3.
Supplier: Abnova Corporation H00009265M01A
Description
This gene encodes a member of the PSCD (pleckstrin homology, Sec7 and coiled-coil domains) family. PSCD family members have identical structural organization that consists of an N-terminal coiled-coil motif, a central Sec7 domain, and a C-terminal pleckstrin homology (PH) domain. The coiled-coil motif is involved in homodimerization, the Sec7 domain contains guanine-nucleotide exchange protein (GEP) activity, and the PH domain interacts with phospholipids and is responsible for association of PSCDs with membranes. Members of this family appear to mediate the regulation of protein sorting and membrane trafficking. This encoded protein is involved in the control of Golgi structure and function, and it may have a physiological role in regulating ADP-ribosylation factor protein 6 (ARF) functions, in addition to acting on ARF1. [provided by RefSeq
Sequence: MNRGINEGGDLPEELLRNLYESIKNEPFKIPEDDGNDLTHTFFNPDREGWLLKLGGRVKTWKRRWFILTDNCLYYFEYTTDKEPRGIIPLENLSIREVEDPRKPNCFELYNPSHKGQVIKACKTEADGRVVEGNHVVYRISAPSPEEKEEWMKSIKASISRDPFYDMLATRKRRIANKKSpecifications
CYTH3 | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
BC008191.1 | |
CYTH3 | |
CYTH3 (AAH08191.1,1 a.a. ∽ 179 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
9265 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Western Blot | |
S1 | |
Mouse monoclonal antibody raised against a full-length recombinant CYTH3. | |
CYTH3 | |
ARNO3/GRP1/PSCD3 | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.