Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-Cytokeratin 14, Clone: KRT14/284, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2450310.2MG
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence
Cytokeratin 14 Monoclonal antibody specifically detects Cytokeratin 14 in Human, Rat, Feline samples. It is validated for Flow Cytometry, Immunofluorescence.Specifications
Cytokeratin 14 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
CK14, CK-14, cytokeratin 14, cytokeratin-14, EBS3, EBS4, K14, keratin 14, keratin 14 (epidermolysis bullosa simplex, Dowling-Meara, Koebner), keratin, type I cytoskeletal 14, Keratin-14, NFJ | |
Mouse | |
50 kDa | |
0.2 mg | |
Cytoskeleton Markers | |
3861 | |
Human, Rat, Feline | |
Purified |
Flow Cytometry, Immunofluorescence | |
KRT14/284 | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
KRT14 | |
Recombinant full-length human KRT14 protein | |
Protein A or G purified | |
RUO | |
Primary | |
Cytokeratin 14 (CK14) belongs to the type I (or A or acidic) subfamily of low molecular weight keratins and exists in combination with keratin 5 (type II or B or basic). CK14 is found in basal cells of squamous epithelia, some glandular epithelia, myoepithelium, and mesothelial cells. Anti-CK14 is useful in differentiating squamous cell carcinomas from poorly differentiated epithelial tumors. Anti-CK14 is one of the specific basal markers for distinguishing between basal and non-basal subtypes of breast carcinomas. Anti-CK14 is also a good marker for differentiation of intraductal from invasive salivary duct carcinoma by the positive staining of basal cells surrounding the in-situ neoplasm as well as for differentiation of benign prostate from prostate carcinoma. Furthermore, this antibody has been useful in separating oncocytic tumors of the kidney from its renal mimics, and in identifying metaplastic carcinomas of the breast. | |
Store at 4C. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction