Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-Cytokeratin 5,6, Clone: D5/16B4, Alexa Fluor 555,

Catalog No. MAB1620AF55 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Alexa Fluor 555
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MAB1620AF55 100 μL Alexa Fluor 555
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1620AF55 Supplier MilliporeSigma™ Supplier No. MAB1620AF555
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Cytokeratin 5,6 clone: D5/16B4 in Human, Mouse, Rat samples, and it is validated for Immunocytochemistry

Keratin, type II cytoskeletal 5 (UniProt P13647; also known as 58kDa cytokeratin, CK-5, Cytokeratin-5, K5, Keratin-5, Type-II keratin Kb5) is encoded by the KRT5 (also known as DDD1, DM-EBS, EBSMCE, K-EBS, MP-EBS, WC-EBS) gene (Gene ID 3852) in human. Cytokeratin-5 (Keratin-5 or K5) belongs to a family of highly conserved epithelial-specific intermediate filament (IF) proteins. Each epithelium is characterized by its unique keratin expression pattern. In interfollicular epidermis, four major keratins are synthesized and co-polymerize in pairs, K5/K14 and K1/KI0, to form filaments. The expression of these two pairs is strictly regulated during normal keratinocyte differentiation. The K5/K14 filament is expressed in proliferating basal cells, while the K1/K10 filament is expressed during the transition to terminal differentiation and at the cessation of cell division. Similar to other IF members, Keratin-5 contains a central rod region composed of alpha helix segments (a.a. 168-203, 223-315, 339-477) that allow two different IF proteins, one type I (acidic) and one type II (basic) keratin, to intertwine into a coiled-coil formation via hydrophobic interaction. Kearain-5 is a type II keratin that pairs with the type I keratin-14 to form IF.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 5,6
Applications Immunocytochemistry
Classification Monoclonal
Clone D5/16B4
Conjugate Alexa Fluor 555
Formulation Purified mouse monoclonal IgG1 antibody conjugate in PBS with 15mg/mL BSA and 0.1% sodium azide.
Gene Accession No. P13647
Gene Symbols KRT5; DDD1; DM-EBS; EBSMCE; K-EBS; MP-EBS; WC-EBS
Host Species Mouse
Immunogen Purified human cytokeratin 5 (Mischke, D., and Wild, G.A. (1987). J. Invest. Dermatol. 88(2):191-197; Wild, G.A., and Mischke, D. (1986). Exp. Cell Res. 162(1):114-126).
Purification Method Protein A Purfied
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NM_000424.3
Target Species Human, Mouse, Rat
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.