Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin, Mouse, Clone: AE1/AE3, Abnova™

Catalog No. 89135446 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-446 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-446 Supplier Abnova Corporation Supplier No. MAB3521
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native Cytokeratin.

Specifications

Antigen Cytokeratin
Applications Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone AE1/AE3
Concentration 70 ug/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native Cytokeratin.
Dilution Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user.
Formulation In phosphate buffer (0.09% sodium azide)
Host Species Mouse
Immunogen Native purified Cytokeratin from human epithelial keratins.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with basic cytokeratins (52-67 kD) and acid cytokeratins (40, 48, 50, 50', 56.5 KDa) according to the Moll nomenclature for CK1-8, 10, 14-16 and 19.
Target Species Bovine, Chicken, Human, Mouse, Rabbit, Rat
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.