Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin, Mouse, Clone: CK-cocktail, Abnova™
Mouse monoclonal antibody raised against native Cytokeratin.
Supplier: Abnova Corporation MAB6194
Specifications
Cytokeratin | |
Monoclonal | |
Unconjugated | |
Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user. | |
Mouse | |
500 μL | |
Primary | |
Store at 4°C. | |
IgG1 κ |
Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
CK-cocktail | |
Mouse monoclonal antibody raised against native Cytokeratin. | |
In PBS, pH 7.4 (1% BSA, 0.05% sodium azide) | |
Native purified from human epidermal keratin. | |
RUO | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction