Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Cytokeratin, HMW, Clone: 34BE12, Azide Free, Novus Biologicals™
SDP

Catalog No. p-5322472 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP23453302 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP23453302 Supplier Novus Biologicals Supplier No. NBP2345330.2 MG

Mouse Monoclonal Antibody

Cytokeratin, HMW Monoclonal antibody specifically detects Cytokeratin, HMW in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.

Specifications

Antigen Cytokeratin, HMW
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 34BE12
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-1.0ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1.0ug/ml, Immunohistochemistry-Paraffin 0.5-1.0ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml, CyTOF-ready
Formulation 10mM PBS - BSA free with No Preservative
Gene Accession No. P04264
Gene Alias CK-2P, cytokeratin 2, Cytokeratin-2P, HUMCYT2A, K2P, K76, keratin 76, keratin, type II cytoskeletal 2 oral, Keratin-76, KRT2Bcytokeratin-2P, KRT2Pkeratin 2p, KRT76, Type-II keratin Kb9
Gene Symbols KRT76
Host Species Mouse
Immunogen Solubilized keratin extract from human stratum corneum
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51350
Test Specificity This antibody recognizes CK1, CK5, CK10 and CK14. In normal epithelia, it stains stratified epithelia, myoepithelial cells and basal cells in the prostate gland and bronchi. This MAb shows no reactivity with hepatocytes, pancreatic acinar cells, proximal renal tubules, or endometrial glands; there is no reactivity with cells derived from simple epithelia. Mesenchymal tumors, lymphomas, melanomas, neural tumors, and neuroendocrine tumors are negative with this antibody. It stains myoepithelial cells and has been shown to be useful in distinguishing prostate adenocarcinoma from benign prostate. This antibody has also been useful in separating benign from malignant intraductal breast proliferations.
Reconstitution Protein A purified
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.