Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin, LMW, Mouse, Clone: AE1, Abnova™

Catalog No. 89109474 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-474 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-474 Supplier Abnova Corporation Supplier No. MAB11310
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Cytokeratin, LMW.

Specifications

Antigen Cytokeratin, LMW
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone AE1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Cytokeratin, LMW.
Dilution Western Blot (0.1 - 1.0 ug/ml) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200 - 1:400) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Gene Alias CK1/EHK1/K1/KRT1A
Gene Symbols KRT1
Host Species Mouse
Immunogen Human epidermal keratin.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3848/3856/3880
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.