Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Dimer, Mouse, Clone: DD2, Abnova™

Catalog No. 89135294 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-294 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-294 Supplier Abnova Corporation Supplier No. MAB2919
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against D-Dimer.

Specifications

Antigen D-Dimer
Applications ELISA, Western Blot
Classification Monoclonal
Clone DD2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against D-Dimer.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Homogenized fibrin clot, human D-dimer or high molecular weight fibrin degradation products.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes D-dimer and high molecular weight fibrin degradation products with different specificities.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.