Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
D-Dimer, Mouse, Clone: DD22, Abnova™
Mouse monoclonal antibody raised against D-Dimer.
Supplier: Abnova Corporation MAB2924
Specifications
D-Dimer | |
Monoclonal | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Mouse | |
200 μg | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
DD22 | |
Mouse monoclonal antibody raised against D-Dimer. | |
In PBS, pH 7.4 (0.09% sodium azide) | |
Homogenized fibrin clot, human D-dimer or high molecular weight fibrin degradation products. | |
RUO | |
This antibody recognizes D-dimer and high molecular weight fibrin degradation products with different specificities. | |
Store at 4°C. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction