Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-dimer (Fibrin Degradation Product), Mouse, Biotin, Clone: DD41, Abnova™

Catalog No. 89135991 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-991 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-991 Supplier Abnova Corporation Supplier No. MAB5487
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against D-dimer (Fibrin Degradation Product).

Specifications

Antigen D-dimer (Fibrin Degradation Product)
Applications Western Blot
Classification Monoclonal
Clone B2654M
Conjugate Biotin
Description Mouse monoclonal antibody raised against D-dimer (Fibrin Degradation Product).
Dilution The optimal working dilution should be determined by the end user.
Formulation In 20mM Tris-HCl, 150mM NaCl, pH 7.5 (0.09% sodium azide)
Host Species Mouse
Immunogen Mixture of D-dimer and high molecular weight fibrin degradation products.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes high molecular weight fibrin degradation products. Recognizes D-dimer in Western Blot under non-reducing conditions. Interacts with beta-chain of D-dimer in Western Blot under reducing conditions. Does not cross-react with fibrinogen and D-monomer.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.