Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DACT3, Mouse, Clone: 2A5, Abnova™

Catalog No. 89110805 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-805 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-805 Supplier Abnova Corporation Supplier No. MAB2064
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant DACT3.

Specifications

Antigen DACT3
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2A5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant DACT3.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene DACT3
Gene Alias MGC15476/RRR1
Gene Symbols DACT3
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 89-202 of human DACT3.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147906
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.