Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAD1, Mouse, Clone: S1, Abnova™

Catalog No. 89104370 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-370 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-370 Supplier Abnova Corporation Supplier No. H00001603M03A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant DAD1.

DAD1, the defender against apoptotic cell death, was initially identified as a negative regulator of programmed cell death in the temperature sensitive tsBN7 cell line. The DAD1 protein disappeared in temperature-sensitive cells following a shift to the nonpermissive temperature, suggesting that loss of the DAD1 protein triggered apoptosis. DAD1 is believed to be a tightly associated subunit of oligosaccharyltransferase both in the intact membrane and in the purified enzyme, thus reflecting the essential nature of N-linked glycosylation in eukaryotes. [provided by RefSeq

Sequence: MSASVVSVISRFLEEYLSSTPQRLKLLDAYLLYILLTGALQFGYCLLVGTFPFNSFLSGFISCVGSFILAVCLRIQINPQNKADFQGISPERAFADFLFASTILHLVVMNFVG

Specifications

Antigen DAD1
Applications ELISA
Classification Monoclonal
Clone S1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant DAD1.
Formulation ascites with no preservative
Gene DAD1
Gene Accession No. BC007403
Gene Alias OST2
Gene Symbols DAD1
Host Species Mouse
Immunogen DAD1 (AAH07403,1 a.a. ∽ 113 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1603
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.