Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAZAP2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106155 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-155 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-155 Supplier Abnova Corporation Supplier No. H00009802A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant DAZAP2.

This gene encodes a proline-rich protein which interacts with the deleted in azoospermia (DAZ) and the deleted in azoospermia-like gene through the DAZ-like repeats. This protein also interacts with the transforming growth factor-beta signaling molecule SARA (Smad anchor for receptor activation), eukaryotic initiation factor 4G, and an E3 ubiquitinase that regulates its stability in splicing factor containing nuclear speckles. The encoded protein may function in various biological and pathological processes including spermatogenesis, cell signaling and transcription regulation, formation of stress granules during translation arrest, RNA splicing, and pathogenesis of multiple myeloma. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: PVGPIYPPGSTVLVEGGYDAGARFGAGATAGNIPPPPPGCPPNAAQLAVMQGANVLVTQRKGNFFMGGSDGGYTIW

Specifications

Antigen DAZAP2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant DAZAP2.
Formulation 50% glycerol
Gene DAZAP2
Gene Accession No. NM_014764
Gene Alias KIAA0058/MGC14319/MGC766/PRTB
Gene Symbols DAZAP2
Host Species Mouse
Immunogen DAZAP2 (NP_055579, 93 a.a. to 168 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9802
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.