Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DBNDD1, Mouse, Clone: 4G4, Abnova™

Catalog No. 89107311 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-311 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-311 Supplier Abnova Corporation Supplier No. H00079007M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant DBNDD1.

Sequence: MAGLPIPPEIVKEAEVPQAALGVPAQGTGDNGHTPVEEEVGGIPVPAPGLLQVTERRQPLSSVSSLEVHFDLLDLTELTDMSDQELAEVFADSDDENLNTESPAGLHPLPRAGYLRSPSWTRTRAEQSHEKQPLGDPERQATVLDTFLTVERPQED

Specifications

Antigen DBNDD1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4G4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant DBNDD1.
Formulation PBS with no preservative; pH 7.4
Gene DBNDD1
Gene Accession No. ENST00000304733
Gene Alias FLJ12582/MGC3101
Gene Symbols DBNDD1
Host Species Mouse
Immunogen DBNDD1 (ENSP00000306407,1 a.a. ∽ 156 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A Purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79007
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.