Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX20, Mouse, Clone: 2A12, Abnova™

Catalog No. 89106489 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-489 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-489 Supplier Abnova Corporation Supplier No. H00011218M04A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant DDX20.

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which has an ATPase activity and is a component of the survival of motor neurons (SMN) complex. This protein interacts directly with SMN, the spinal muscular atrophy gene product, and may play a catalytic role in the function of the SMN complex on RNPs. [provided by RefSeq

Sequence: MAAAFEASGALAAVATAMPAEHVAVQVPAPEPTPGPVRILRTAQDLSSPRTRTGDVLLAEPADFESLLLSRPVLEGLRAAGFERPSPVQLKAIPLGRCGLDLIVQAKSGTGKTCVFSTIALDSLVLENLSTQILILAPTREIAVQIHSVITAIGIKMEGLECHVFIGGTPLSQDKTRLKKCHIAVGSPGRIKQLIELDYLNPGSIRLFILDEADKLLEEGSFQEQINWIYSSLPASKQMLAVSATYPEFLANALTKYMRDPTFVRLNSSDPSLIGLKQYYKVVNSYPLAHKVFEEKTQHLQELFSRIPFNQALVFSNLHSRAQHLADILSSKGFPAECISGNMNQNQRLDAMAKLKHFHCRVLISTDLTSRGIDAEKVNLVVNLDVPLDWETYMHRIGRAGRFGTLGLTVTYCCRGEEGNMMMRIAQKCNINLLPLPDPIPSGLMEECVDWDVEVKAAVHTYGIASVPNQPLKKQIQKIERTLQIQKAHGDHMASSRNNSVSGLSVKSKNNTKQKLPVKSHSECGIIEKATSPKELGCDRQSEEQMKNSVQTPVENSTNSQHQVKEALPVSLPQIPCLSSFKIHQPYTLTFAELVEDYEHYIKEGLEKPVEIIRHYTGPGDQTVNPQNGFVRNKVTEQRVPVLASSSQSGDSESDSDSYSSRTSSQSKGNKSYLEGSSDNQLKDSESTPVDDRISLEQPPNGSDTPNPEKYQESPGIQMKTRLKEGASQRAKQSRRNLPRRSSFRLQTEAQEDDWYDCHREIRLSFSDTYQDYEEYWRAYYRAWQEYYAAASHSYYWNAQRHPSWMAAYHMNTIYLQEMMHSNQ

Specifications

Antigen DDX20
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2A12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant DDX20.
Formulation ascites with no preservative
Gene DDX20
Gene Accession No. BC034953
Gene Alias DKFZp434H052/DP103/GEMIN3
Gene Symbols DDX20
Host Species Mouse
Immunogen DDX20 (AAH34953,1 a.a. ∽ 824 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11218
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.