Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DEF6, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106833 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-833 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-833 Supplier Abnova Corporation Supplier No. H00050619B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human DEF6 protein.

DEF6, or IBP, is a guanine nucleotide exchange factor (GEF) for RAC (MIM 602048) and CDC42 (MIM 116952) that is highly expressed in B and T cells (Gupta et al., 2003 [PubMed 12923183]).[supplied by OMIM

Sequence: MALRKELLKSIWYAFTALDVEKSGKVSKSQLKVLSHNLYTVLHIPHDPVALEEHFRDDDDGPVSSQGYMPYLNKYILDKVEEGAFVKEHFDELCWTLTAKKNYRADSNGNSMLSNQDAFRLWCLFNFLSEDKYPLIMVPDEVEYLLKKVLSSMSLEVSLGELEELLAQEAQVAQTTGGLSVWQFLELFNSGRCLRGVGRDTLSMAIHEVYQELIQDVLKQGYLWKRGHLRRNWAERWFQLQPSCLCYFGSEECKEKRGIIPLDAHCCVEVLPDRDGKRCMFCVKTATRTYEMSASDTRQRQEWTAAIQMAIRLQAEGKTSLHKDLKQKRREQREQRERRRAAKEEELLRLQQLQEEKERKLQELELLQEAQRQAERLLQEEEERRRSQHRELQQALEGQLREAEQARASMQAEMELKEEEAARQRQRIKELEEMQQRLQEALQLEVKARRDEESVRIAQTRLLEEEEEKLKQLMQLKEEQERYIERAQQEKEELQQEMAQQSRSLQQAQQQLEEVRQNRQRADEDVEAAQRKLRQASTNVKHWNVQMNRLMHPIEPGDKRPVTSSSFSGFQPPLLAHRDSSLKRLTRWGSQGNRTPSPNSNEQQKSLNGGDEAPAPASTPQEDKLDPAPEN

Specifications

Antigen DEF6
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human DEF6 protein.
Formulation PBS with no preservative; pH 7.4
Gene DEF6
Gene Accession No. NM_022047.2
Gene Alias IBP/SLAT
Gene Symbols DEF6
Host Species Mouse
Immunogen DEF6 (NP_071330.2,1 a.a. ∽ 631 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50619
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.