Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DEFA1, Mouse, Clone: B539M, Abnova™

Catalog No. 89112920 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-920 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-920 Supplier Abnova Corporation Supplier No. MAB6523

Mouse monoclonal antibody raised against synthetic peptide of DEFA1.

Defensins are a family of microbicidal and cytotoxic peptides thought to be involved in host defense. They are abundant in the granules of neutrophils and also found in the epithelia of mucosal surfaces such as those of the intestine, respiratory tract, urinary tract, and vagina. Members of the defensin family are highly similar in protein sequence and distinguished by a conserved cysteine motif. Several alpha defensin genes appear to be clustered on chromosome 8. The protein encoded by this gene, defensin, alpha 1, is found in the microbicidal granules of neutrophils and likely plays a role in phagocyte-mediated host defense. It differs from defensin, alpha 3 by only one amino acid. [provided by RefSeq

Specifications

Antigen DEFA1
Applications ELISA, Western Blot
Classification Monoclonal
Clone B539M
Conjugate Unconjugated
Description defensin, alpha 1
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene DEFA1
Gene Alias DEF1, DEFA2, HNP-1, HP-1, MGC138393, MRS
Gene Symbols DEFA1
Host Species Mouse
Immunogen A synthetic peptide (conjugated with Hsp70) corresponding to human DEFA1.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1667
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.