Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Defensin, Mouse, Clone: D1H5, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of Defensin.
Supplier: Abnova Corporation MAB2931
Specifications
Defensin | |
Monoclonal | |
Unconjugated | |
Mouse | |
200 μg | |
Primary | |
Store at 4°C. | |
IgG1 |
ELISA, Western Blot | |
D1H5 | |
In PBS, pH 7.4 (0.09% sodium azide) | |
A synthetic peptide (conjugated with Hsp70) corresponding to human HD1. | |
RUO | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction