Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Defensin, Mouse, Clone: D2C6, Abnova™

Catalog No. 89135307 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-307 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-307 Supplier Abnova Corporation Supplier No. MAB2932

Mouse monoclonal antibody raised against recombinant Defensin.

Specifications

Antigen Defensin
Applications ELISA, Western Blot
Classification Monoclonal
Clone D2C6
Conjugate Unconjugated
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Recombinant hsp70 fusion protein corresponding to human HD2.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.