Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DEGS1, Mouse, Clone: 3F9, Abnova™

Catalog No. 89105889 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-889 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-889 Supplier Abnova Corporation Supplier No. H00008560M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant DEGS1.

This gene encodes a member of the membrane fatty acid desaturase family which is responsible for inserting double bonds into specific positions in fatty acids. This protein contains three His-containing consensus motifs that are characteristic of a group of membrane fatty acid desaturases. It is predicted to be a multiple membrane-spanning protein localized to the endoplasmic reticulum. Overexpression of this gene inhibited biosynthesis of the EGF receptor, suggesting a possible role of a fatty acid desaturase in regulating biosynthetic processing of the EGF receptor. Two splice variants have been identified. [provided by RefSeq

Sequence: SGHFIAEHYMFLKGHETYSYYGPLNLLTFNVGYHNEHHDFPNIPGKSLPLVRKIAAEYYDNLPHYNSWIKVLYDFVMDDTISPYSRMKRHQKGEMVLE

Specifications

Antigen DEGS1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3F9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant DEGS1.
Formulation ascites with no preservative
Gene DEGS1
Gene Accession No. NM_003676
Gene Alias DEGS/DES1/Des-1/FADS7/MGC5079/MIG15/MLD
Gene Symbols DEGS1
Host Species Mouse
Immunogen DEGS1 (NP_003667,226 a.a. ∽ 323 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8560
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.