Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-Derlin-1, Clone: 8G11.2,

Catalog No. MABS1154 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS1154 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1154 Supplier MilliporeSigma™ Supplier No. MABS1154
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Derlin-1 clone: 8G11.2 in Human, Mouse samples, and it is validated for Immunocytochemistry, Western Blotting

Derlin-1 (UniProt Q9BUN8; also known as Degradation in endoplasmic reticulum protein 1, Der1-like protein 1, DERtrin-1) is encoded by the DERL1 (also known as DER1) gene (ORF UNQ243/PRO276; Gene ID 79139) in human. Derlins are integral ER membrane proteins that form homo-oligomers and heteromultimeric complexes with membrane-bound machinery, including the E3 ubiquitin ligase Hrd1 and its binding partner SEL1L, involved in ER-associated degradation (ERAD). Before ERAD substrates can be processed by the cytoplasmic ubiquitin-proteasome system, they need to be translocated across the ER membrane through an AAA ATPase p97/VCP-driven process. Derlins are inactive members of the rhomboid family of intramembrane proteases that mediate the recruitment of AAA ATPase p97/VCP through interaction via their C-terminal SHP boxes. Derlin-1 is proposed to contain four transmembrane segments (a.a. 23-43, 60-80, 106-126, 155-175), three cytoplasmic domains (a.a. 23-43, 60-80, 106-126, 155-175), and two lumenol (a.a. 44-59 & 127-154) domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Derlin-1
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 8G11.2
Formulation Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q9BUN8
Gene Symbols DERL1; DER1; UNQ243/PRO276
Host Species Mouse
Immunogen GST-tagged recombinant human derlin-1 C-terminal cytoplasmic fragment.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001128143
Target Species Human, Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.