Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DES, Mouse, Clone: D33, Abnova™

Catalog No. 89111392 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-392 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-392 Supplier Abnova Corporation Supplier No. MAB3396
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against DES.

This gene encodes a muscle-specific class III intermediate filament. Homopolymers of this protein form a stable intracytoplasmic filamentous network connecting myofibrils to each other and to the plasma membrane. Mutations in this gene are associated with desmin-related myopathy, a familial cardiac and skeletal myopathy (CSM), and with distal myopathies. [provided by RefSeq

Specifications

Antigen DES
Applications Immunofluorescence, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone D33
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against DES.
Dilution Immunohistochemistry (1:100) The optimal working dilution should be determined by the end user.
Formulation In 150mM PBS (0.09% sodium azide)
Gene DES
Gene Alias CMD1I/CSM1/CSM2/FLJ12025/FLJ39719/FLJ41013/FLJ41793
Gene Symbols DES
Host Species Mouse
Immunogen Native purified human DES.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1674
Test Specificity This antibody is highly reactive with desmin. On immunoblots, only the 52 KDa desmin band is stained. This antibody stains muscle cells in human tissues. No detectable reactivity with GFAP, keratin, vimentin or neurofilament.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.