Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmin, Mouse, Clone: DE-U-10, Abnova™

Catalog No. 89134962 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-962 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-962 Supplier Abnova Corporation Supplier No. MAB1452
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length native pig desmin.

Specifications

Antigen Desmin
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone DE-U-10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length native pig desmin.
Dilution Immunohistochemistry (2-4 ug/mL) Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 1.2% sodium acetate (2mg BSA, 0.01mg sodium azide)
Gene LOC396725
Gene Symbols LOC396725
Host Species Mouse
Immunogen Native purified desmin from pig stomach.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 396725
Target Species Human, Mouse, Rabbit, Rat
Content And Storage Store at -20°C. After reconstitution with 1mL of 1.2% sodium acetate or neutral PBS to a concentration of 100μg/mL, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.