Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Digoxin, Mouse, Clone: D8-9, Abnova™
Mouse monoclonal antibody raised against Digoxin.
Supplier: Abnova Corporation MAB8237
Specifications
Digoxin | |
Monoclonal | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Mouse | |
100 μg | |
Primary | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
ELISA | |
D8-9 | |
Mouse monoclonal antibody raised against Digoxin. | |
In PBS | |
Digoxin conjugated with KLH. | |
RUO | |
This antibody specifically recognize digoxin. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction