Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DLG4, Mouse, Clone: 6G6-1C9, Abnova™

Catalog No. 89113413 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-413 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 89-113-413 Supplier Abnova Corporation Supplier No. MAB8280
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse monoclonal antibody raised against recombinant DLG4.

Specifications

Antigen DLG4
Applications Western Blot
Classification Monoclonal
Clone 6G6-1C9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant DLG4.
Dilution Western Blot (4 ug/mL) Immunocytochemistry (3-15 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 0.09% sodium azide)
Gene Dlg4
Gene Alias Dlgh4/PSD95/Sap90
Gene Symbols Dlg4
Host Species Mouse
Immunogen Recombinant protein corresponding to rat DLG4.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29495
Test Specificity This antibody recognizes mouse, rat, and bovine PSD95.
Target Species Bovine, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.