Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DLG4, Mouse, Clone: 7E3-1B8, Abnova™

Catalog No. 89113414 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-414 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-414 Supplier Abnova Corporation Supplier No. MAB8282
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant DLG4.

Specifications

Antigen DLG4
Applications Western Blot
Classification Monoclonal
Clone 7E3-1B8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant DLG4.
Dilution Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 0.09% sodium azide)
Gene Dlg4
Gene Alias Dlgh4/PSD95/Sap90
Gene Symbols Dlg4
Host Species Mouse
Immunogen Recombinant protein corresponding to rat DLG4.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29495
Target Species Bovine, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.