Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DLG5, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106033 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-033 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-033 Supplier Abnova Corporation Supplier No. H00009231A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant DLG5.

This gene encodes a member of the family of discs large (DLG) homologs, a subset of the membrane-associated guanylate kinase (MAGUK) superfamily. The MAGUK proteins are composed of a catalytically inactive guanylate kinase domain, in addition to PDZ and SH3 domains, and are thought to function as scaffolding molecules at sites of cell-cell contact. The protein encoded by this gene localizes to the plasma membrane and cytoplasm, and interacts with components of adherens junctions and the cytoskeleton. It is proposed to function in the transmission of extracellular signals to the cytoskeleton and in the maintenance of epithelial cell structure. Alternative splice variants have been described but their biological nature has not been determined. [provided by RefSeq

Sequence: LDIAPHAIERLHHMHIYPIVIFIHYKSAKHIKEQRDPIYLRDKVTQRHSKEQFEAAQKLEQEYSRYFTGVIQGGALSSICTQILAMVNQEQNKVLWIPACPL

Specifications

Antigen DLG5
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant DLG5.
Formulation 50% glycerol
Gene DLG5
Gene Accession No. NM_004747
Gene Alias KIAA0583/LP-DLG/P-DLG5/PDLG
Gene Symbols DLG5
Host Species Mouse
Immunogen DLG5 (NP_004738, 1708 a.a. to 1809 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9231
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.