Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DMC1, Mouse, Clone: 1D12/4, Abnova™

Catalog No. 89110949 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-949 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-949 Supplier Abnova Corporation Supplier No. MAB2349
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant DMC1.

The protein encoded by this gene is essential for meiotic homologous recombination. Genetic recombination in meiosis plays an important role in generating diversity of genetic information. The product of this gene is structurally and evolutionary related to the products of the yeast RAD51 and E. coli RecA genes. Alternative splice variants of this gene have been described but their full-length nature has not been determined. [provided by RefSeq

Specifications

Antigen DMC1
Applications Western Blot
Classification Monoclonal
Clone 1D12/4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant DMC1.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.09% sodium azide
Gene DMC1
Gene Alias DMC1H/HsLim15/LIM15/MGC150472/MGC150473/dJ199H16.1
Gene Symbols DMC1
Host Species Mouse
Immunogen Recombinant His fusion protein corresponding to human DMC1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11144
Test Specificity This antibody is specific to DMC-1.
Target Species Human, Mouse
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.