Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DMPO, Mouse, APC, Clone: N1664A, Abnova™

Catalog No. 89134868 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-868 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-868 Supplier Abnova Corporation Supplier No. MAB11436
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against DMPO.

Specifications

Antigen DMPO
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone N1664A
Conjugate APC
Description Mouse monoclonal antibody raised against DMPO.
Dilution Enzyme-Linked Immunoabsorbent Assay (1-10 ug/mL) Immunocytochemistry (10 ug/mL) Immunoprecipitation (25 ug) The optimal working dilution should be determined by the end user.
Formulation In TBS, pH 7.4 (50% glycerol, 0.05% sodium azide)
Host Species Mouse
Immunogen 5, 5-dimethyl-2-(8-octanoic acid)-1-pyrrolone-N-oxide conjugated with ovalbumin.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes DMPO, DMPO-octanoic acid, DMPO-protein adducts and DMPO-DNA adducts. Does not cross react with non-adducted proteins or DNA.
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.