Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAJA1, Mouse, Clone: KA2A5.6, Abnova™

Catalog No. 89111575 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-575 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-575 Supplier Abnova Corporation Supplier No. MAB3738

Mouse monoclonal antibody raised against partial recombinant DNAJA1.

Specifications

Antigen DNAJA1
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone KA2A5.6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant DNAJA1.
Dilution Immunoprecipitation (2 ug/mg protein lysate) Immunohistochemistry (1-2 ug/mL) Western Blot (0.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.4 (0.09% sodium azide)
Gene DNAJA1
Gene Alias DJ-2/DjA1/HDJ2/HSDJ/HSJ2/HSPF4/hDJ-2
Gene Symbols DNAJA1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-179 of human DNAJA1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3301
Test Specificity Clone KA2A5.6 recognizes a 44-47 KDa protein identified as HDJ-2. This antibody is highly specific to HDJ-2 and shows no cross-reaction with HDJ-1. HDJ-2 has a prenylated C-terminus and is not inducible upon heat shock. It associates with a 53KDa protein of unknown identity.
Target Species Bacteria, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.