Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNMT1, Mouse, ATTO594, Clone: 4G11-C7, Abnova™

Catalog No. 89109902 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-902 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-902 Supplier Abnova Corporation Supplier No. MAB11768
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant DNMT1.

DNA (cytosine-5-)-methyltransferase 1 has a role in the establishment and regulation of tissue-specific patterns of methylated cytosine residues. Aberrant methylation patterns are associated with certain human tumors and developmental abnormalities. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen DNMT1
Applications ChIP Assay, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4G11-C7
Conjugate PE/ATTO 594
Description Mouse monoclonal antibody raised against partial recombinant DNMT1.
Dilution Western Blot (2-4 ug/mL) Immunohistochemistry (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Gene DNMT1
Gene Alias AIM/CXXC9/DNMT/FLJ16293/MCMT/MGC104992
Gene Symbols DNMT1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids of 620-950 of human DNMT1.
Purification Method Protein G Purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1786
Test Specificity The antibody will cross-react with mouse DNMT1.
Target Species Human
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.