Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dnmt3a, Mouse, Clone: 64B1446, Abnova™

Catalog No. 89107851 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-851 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-851 Supplier Abnova Corporation Supplier No. MAB0080
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant Dnmt3a.

Specifications

Antigen Dnmt3a
Applications ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 64B1446
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant Dnmt3a.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Gene Dnmt3a
Gene Alias AU015806/W91664
Gene Symbols Dnmt3a
Host Species Mouse
Immunogen Recombinant His fusion protein corresponding to full length mouse Dnmt3a.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13435
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.