Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DOCK4, Mouse, Clone: 3E7, Abnova™

Catalog No. 89106145 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-145 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-145 Supplier Abnova Corporation Supplier No. H00009732M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant DOCK4.

This gene is a member of the dedicator of cytokinesis (DOCK) family and encodes a protein with a DHR-1 (CZH-1) domain, a DHR-2 (CZH-2) domain and an SH3 domain. This membrane-associated, cytoplasmic protein functions as a guanine nucleotide exchange factor and is involved in regulation of adherens junctions between cells. Mutations in this gene have been associated with ovarian, prostate, glioma, and colorectal cancers. Alternatively spliced variants which encode different protein isoforms have been described, but only one has been fully characterized. [provided by RefSeq

Sequence: NQVNEQSAPLPVPVPVPVPSYGGEEPVRKESKTPPPYSVYERTLRRPVPLPHSLSIPVTSEPPALPPKPLAARSSHLENGARRTDPGPRPRPLPRKVSQL

Specifications

Antigen DOCK4
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3E7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant DOCK4.
Formulation ascites with no preservative
Gene DOCK4
Gene Accession No. NM_014705
Gene Alias FLJ34238/KIAA0716/MGC134911/MGC134912
Gene Symbols DOCK4
Host Species Mouse
Immunogen DOCK4 (NP_055520,1867 a.a. ∽ 1966 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9732
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.