Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-DOCK8, Clone: 10C3.2,

Catalog No. MABN353MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN353MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN353MI Supplier MilliporeSigma™ Supplier No. MABN353
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects DOCK8 clone: 10C3.2 in Human samples, and it is validated for Flow Cytometry, Western Blotting

Dedicator of cytokinesis protein 8 (also known as 1200017A24Rik or Epididymis luminal protein 205) is encoded by the DOCK8 gene (also known as HEL-205, MRD2, ZIR8) in human (NCBI Gene ID 81704). DOCK8 is a cytosolic scaffolding protein involved in cytoskeletal rearrangement and cell migration. DOCK8 deficiency due to genetic mutations result in combined immune deficiency, historically diagnosed as an atypical form of hyper-IgE syndrome (HIES) or autosomal recessive HIES, which remains to be the focus of ongoing DOCK8 research activities. (PMIDn20864884). Clone 10C3.2 detects an internal epitope within the C-terminal half of the DOCK8 protein (UniProt Q8NF50).
TRUSTED_SUSTAINABILITY

Specifications

Antigen DOCK8
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 10C3.2
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q8NF50
Gene Symbols DOCK8
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to human DOCK8.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001177387
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.