Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPP4, Mouse, Clone: 202.36, Abnova™

Catalog No. 89112211 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-211 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-211 Supplier Abnova Corporation Supplier No. MAB5299
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native DPP4.

The protein encoded by this gene is identical to adenosine deaminase complexing protein-2, and to the T-cell activation antigen CD26. It is an intrinsic membrane glycoprotein and a serine exopeptidase that cleaves X-proline dipeptides from the N-terminus of polypeptides. [provided by RefSeq

Specifications

Antigen DPP4
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone 202.36
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native DPP4.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene DPP4
Gene Alias ADABP/ADCP2/CD26/DPPIV/TP103
Gene Symbols DPP4
Host Species Mouse
Immunogen Native purified DPP4 from T cell clone.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1803
Test Specificity 202.36 reacts with human CD26 (Dipeptidyl peptidase IV), a 110 kD protein which occurs in the cell membrane of T lymphocyte and is upregulated after activation. CD26 is also present on endothelial cells. The mAb was clustered at the VI WLDA.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.