Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dpp4, Mouse, Clone: 5E8, Abnova™

Catalog No. 89112343 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-343 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-343 Supplier Abnova Corporation Supplier No. MAB5649
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native Dpp4.

Specifications

Antigen Dpp4
Applications ELISA, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 5E8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native Dpp4.
Dilution Recommend dilution at 1:10 The optimal working dilution should be determined by the end user.
Formulation In PBS (0.1% BSA, 0.02% sodium azide)
Gene Dpp4
Gene Alias DPPIV
Gene Symbols Dpp4
Host Species Mouse
Immunogen Native purified rat liver membrane extracts Dpp4.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25253
Test Specificity Dipeptidyl peptidase IV (DPP IV) is widely distributed in a number of mammalian tissues and is suggested to play an important role in various kinds of biological processes. DPP IV (CD26) is a serine-type protease that removes the amino-terminal dipeptide from peptide substrate provided that the penultimate amino acid residue is proline or alanine. DPP IV plays an important role in the reclamation of peptide nitrogen from larger peptides. The monoclonal antibody 5E8 reacts with DPP IV present on the apical surface of epithelial cells in the pancreas, small intestine, colon, and bile duct. Furthermore antibody 5E8 reacts with DPP IV on the laminar portions of the proximal renal tubule cells, and, weakly, on the glomeruli.
Target Species Rat
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.