Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Drd1a, Mouse, Clone: SG2-D1a, Abnova™

Catalog No. 89110925 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-925 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-925 Supplier Abnova Corporation Supplier No. MAB2314
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant Drd1a.

Specifications

Antigen Drd1a
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone SG2-D1a
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant Drd1a.
Dilution The optimal working dilution should be determined by the end user.
Formulation In Tris-glycine, 150mM NaCl (0.05% sodium azide)
Gene Drd1a
Gene Alias D1a/Drd-1/Drd1
Gene Symbols Drd1a
Host Species Mouse
Immunogen Recombinant protein corresponding to C-terminus of rat Drd1a.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24316
Test Specificity This antibody reacts with Dopamine Receptor D1A. There is no cross- reactivity with D1B (D5) receptor.
Target Species Mouse, Rat
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.