Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DSC3, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104398 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-398 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-398 Supplier Abnova Corporation Supplier No. H00001825A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant DSC3.

The protein encoded by this gene is a calcium-dependent glycoprotein that is a member of the desmocollin subfamily of the cadherin superfamily. These desmosomal family members, along with the desmogleins, are found primarily in epithelial cells where they constitute the adhesive proteins of the desmosome cell-cell junction and are required for cell adhesion and desmosome formation. The desmosomal family members are arranged in two clusters on chromosome 18, occupying less than 650kb combined. Alternative splicing results in two transcript variants encoding distinct isoforms. [provided by RefSeq

Sequence: CKPKMGYTDILAVDPDEPVHGAPFYFSLPNTSPEISRLWSLTKVNDTAARLSYQKNAGFQEYTIPITVKDRAGQAATKLLRVNLCECTHPTQCRATSRST

Specifications

Antigen DSC3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant DSC3.
Formulation 50% glycerol
Gene DSC3
Gene Accession No. NM_001941
Gene Alias CDHF3/DSC/DSC1/DSC2/DSC4/HT-CP
Gene Symbols DSC3
Host Species Mouse
Immunogen DSC3 (NP_001932, 585 a.a. to 684 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1825
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.