Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DSE (B01P), Mouse anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89106799 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-799 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-799 Supplier Abnova Corporation Supplier No. H00029940B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human DSE protein.

The protein encoded by this gene is a tumor-rejection antigen. This antigen possesses tumor epitopes capable of inducing HLA-A24-restricted and tumor-specific cytotoxic T lymphocytes in cancer patients and may be useful for specific immunotherapy. This gene product is localized to the endoplasmic reticulum. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq

Sequence: MRTHTRGAPSVFFIYLLCFVSAYITDENPEVMIPFTNANYDSHPMLYFSRAEVAELQLRAASSHEHIAARLTEAVHTMLSSPLEYLPPWDPKDYSARWNEIFGNNLGALAMFCVLYPENIEARDMAKDYMERMAAQPSWLVKDAPWDEVPLAHSLVGFATAYDFLYNYLSKTQQEKFLEVIANASGYMYETSYRRGWGFQYLHNHQPTNCMALLTGSLVLMNQGYLQEAYLWTKQVLTIMEKSLVLLREVTDGSLYEGVAYGSYTTRSLFQYMFLVQRHFNINHFGHPWLKQHFAFMYRTILPGFQRTVAIADSNYNWFYGPESQLVFLDKFVMRNGSGNWLADQIRRNRVVEGPGTPSKGQRWCTLHTEFLWYDGSLKSVPPPDFGTPTLHYFEDWGVVTYGSALPAEINRSFLSFKSGKLGGRAIYDIVHRNKYKDWIKGWRNFNAGHEHPDQNSFTFAPNGVPFITEALYGPKYTFFNNVLMFSPAVSKSCFSPWVGQVTEDCSSKWSKYKHDLAASCQGRVVAAEEKNGVVFIRGEGVGAYNPQLNLKNVQRNLILLHPQLLLLVDQIHLGEESPLETAASFFHNVDVPFEETVVDGVHGAFIRQRDGLYKMYWMDDTGYSEKATFASVTYPRGYPYNGTNYVNVTMHLRSPITRAAYLFIGPSIDVQSFTVHGDSQQLDVFIATSKHAYATYLWTGEATGQSAFAQVIADRHKILFDRNSAIKSSIVPEVKDYAAIVEQNLQHFKPVFQLLEKQILSRVRNTASFRKTAERLLRFSDKRQTEEAIDRIFAISQQQQQQSKSKKNRRAGKRYKFVDAVPDIFAQIEVNEKKIRQKAQILAQKELPIDEDEEMKDLLDFADVTYEKHKNGGLIKGRFGQARMVTTTHSRAPSLSASYTRLFLILNIAIFFVMLAMQLTYFQRAQSLHGQRCLYAVLLIDSCILLWLYSSCSQSQC

Specifications

Antigen DSE
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human DSE protein.
Formulation PBS with no preservative; pH 7.4
Gene DSE
Gene Accession No. NM_013352.1
Gene Alias DSEPI/SART2
Gene Symbols DSE
Host Species Mouse
Immunogen DSE (NP_037484.1,1 a.a. ∽ 958 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29940
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.