Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DSG1, Mouse, Clone: CC49, Abnova™

Catalog No. 89110289 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-289 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-289 Supplier Abnova Corporation Supplier No. MAB12146
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against human DSG1.

Desmosomes are cell-cell junctions between epithelial, myocardial and certain other cell types. Desmoglein 1 is a calcium-binding transmembrane glycoprotein component of desmosomes in vertebrate epithelial cells. Currently, three desmoglein subfamily members have been identified and all are members of the cadherin cell adhesion molecule superfamily. These desmoglein gene family members are located in a cluster on chromosome 18. The protein encoded by this gene has been identified as the autoantigen of the autoimmune skin blistering disease pemphigus foliaceus. [provided by RefSeq

Specifications

Antigen DSG1
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone CC49
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human DSG1.
Dilution Immunohistochemistry (0.5-1 ug/mL) Western blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Gene DSG1
Gene Alias CDHF4/DG1/DSG
Gene Symbols DSG1
Host Species Mouse
Immunogen Purified human DSG1 protein.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1828
Target Species Human
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.