Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-dUTPase, Clone: 6A6.2,

Catalog No. MABC608MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC608MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC608MI Supplier MilliporeSigma™ Supplier No. MABC608
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects dUTPase clone: 6A6.2 in Human samples, and it is validated for Immunocytochemistry, Western Blotting

Deoxyuridine 5'-triphosphate nucleotidohydrolase (EC 3.6.1.23; also known as dUTPase, dUTP nucleotidohydrolase, or dUTP pyrophosphatase) is encoded by the DUT gene (Entrez Gene ID 1854) in human. dUTPase catalyzes the hydrolysis of dUTP to generate dUMP. This reaction serves to provide dUMP of thymine nucleotides synthesis needed for DNA replication. dUTP hydrolysis by dUTPase also prevents increased incorporation of uracil into DNA, which induces extensive excision repair mediated by uracil glycosylase. This repair process, resulting in the removal and reincorporation of dUTP, is self-defeating and leads to DNA fragmentation and cell death. The DUT gene encodes both mitochondrial and nuclear dATPase (DUT-M and DUT-N, respectively) via alternative splicing. DUT-N (164 aa, 17.7kDa) corresponds to DUT-M (252 aa, 26.6kDa) with the first 93 amino acid sequence replaced by MPCSE. Anti-dUTPase, clone 6A6.2, is a purified mouse monoclonal antibody. A GST fusion protein containing the full-lenght human DUT-N (UniProt P33316) sequence was used as the immunogen. The binding epitope of clone 6A6.2 is not yet mapped.
TRUSTED_SUSTAINABILITY

Specifications

Antigen dUTPase
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 6A6.2
Formulation Purified mouse monoclonal IgG2bκ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P33316
Gene Symbols DUT
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human dUTPase.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_001020419
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.