Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUX4, Mouse, Biotin, Clone: P2B1, Abnova™

Catalog No. 89109603 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-603 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-603 Supplier Abnova Corporation Supplier No. MAB11453
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant partial DUX4.

This gene is located within a D4Z4 repeat array in the subtelomeric region of chromosome 4q. The D4Z4 repeat is polymorphic in length; a similar D4Z4 repeat array has been identified on chromosome 10. Each D4Z4 repeat unit has an open reading frame (named DUX4) that contains two homeoboxes; the repeat-array and ORF is conserved in other mammals. There was no evidence for transcription from standard cDNA libraries however RTPCR and in-vitro expression experiments indicate that the ORF is transcribed and the encoded protein has been reported to function as a transcriptional activator of paired-like homeodomain transcription factor 1 (PITX1; GeneID 5307). Contraction of the microsatellite repeat causes autosomal dominant facioscapulohumeral muscular dystrophy (FSHD). [provided by RefSeq

Specifications

Antigen DUX4
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone P2B1
Conjugate Biotin
Description Mouse monoclonal antibody raised against recombinant partial DUX4.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene DUX4
Gene Alias DUX10
Gene Symbols DUX4
Host Species Mouse
Immunogen Recombinant GST fusion protein corresponding to a 76 amino acid fragment at C-terminus of human DUX4.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22947
Test Specificity This antibody recognizes human DUX4. It does not cross-react with DUX4c.
Target Species Human
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.