Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DYKDDDDK tag, Mouse, Clone: G191, Abnova™

Catalog No. 89136431 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-431 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-431 Supplier Abnova Corporation Supplier No. MAB8183
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptides of DYKDDDDK tag.

Specifications

Antigen DYKDDDDK tag
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone G191
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptides of DYKDDDDK tag.
Dilution Western Blot (1:1000-1:4000) Immunoprecipitation (1:200-1:400) The optimal working dilution should be determined by the end user.
Formulation In 20mM PBS, 0.15M NaCl, pH 7.2 (0.01% sodium azide)
Host Species Mouse
Immunogen A synthetic peptide (conjugated with KLH) corresponding to DYKDDDDK tag.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes over-expressed proteins containing Flag epitope tag (D-tag) fused to either amino- or carboxy-termini of targeted proteins in transfected mammalian cells.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.