Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-Dynein, Clone: 74.1, Alexa Fluor 555,

Catalog No. MAB1618AF55 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Alexa Fluor 555
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MAB1618AF55 100 μL Alexa Fluor 555
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1618AF55 Supplier MilliporeSigma™ Supplier No. MAB1618AF555
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Dynein clone: 74.1 in Bovine, Drosophila, Fish, Human, Mouse, Rat, Sheep, Xenopus samples, and it is validated for Immunocytochemistry

Cytoplasmic dynein 1 intermediate chain 1 (UniProt Q29RQ3; also known as Cytoplasmic dynein intermediate chain 1, Dynein intermediate chain 1, cytosolic, DH IC-1) and Cytoplasmic dynein 1 intermediate chain 2 (UniProt Q0III3; also known as Cytoplasmic dynein intermediate chain 2, Dynein intermediate chain 2, cytosolic, DH IC-2) are encoded by the DYNC1I1 gene (Gene ID 613724) and DYNC1I2 gene (Gene ID 526329) in bovine, respectively. Cytoplasmic dynein complexes are microtubules-associated retrograde transport motors. Cytoplasmic dynein 1 is the more abundant motor in cells, while cytoplasmic dynein 2 takes part in intraflagellar transport. Cytoplasmic dynein 1 is a multisubunit complex of ∼1.5 MDa composed of a homodimer of heavy chains (encoded by DYNC1H1), two intermediate chains (encoded by DYNC1I1 and DYNC1I2), light-intermediate chains (encoded by DYNC1LI1, DYNC1LI2) and light chains (encoded by DYNLT1, DYNLT3, DYNLRB1, DYNLRB2, DYNLL1, DYNLLl2). The heavy chain homodimer acts as the dynein complex core and binds microtubules to enable ATP-dependent cytoplasmic dynein movement. The other dynein subunits associate as homodimers and play regulatory roles in maintaining the complex stability, modulating its activity, and mediating its interaction with accessory and cargo proteins. The intermediate chain proteins (IC1 & IC2) are involved in cargo binding and specificity, either alone or via interaction with the dynactin complex. IC1 and IC2 share 69% protein identity. They interact with the dynein light chains and the p150 subunit of dynactin at the N-terminus and with the heavy chains through WD40 repeats at the intermediate chain C-terminus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dynein
Applications Immunocytochemistry
Classification Monoclonal
Clone 74.1
Conjugate Alexa Fluor 555
Formulation Purified mouse monoclonal IgG2b antibody conjugate in PBS with 15mg/mL BSA and 0.1% sodium azide.
Gene Accession No. Q29RQ3; Q0III3
Gene Symbols DYNC1I1; DYNC1I2
Host Species Mouse
Immunogen Bovine brain cytoplasmic dynein.
Purification Method Protein A Purfied
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NM_012144.2
Target Species Bovine, Drosophila, Fish, Human, Mouse, Rat, Sheep, Xenopus species
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.