Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E2F3, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104412 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-412 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-412 Supplier Abnova Corporation Supplier No. H00001871A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length recombinant E2F3.

The protein encoded by this gene is a member of the E2F family of transcription factors. The E2F family plays a crucial role in the control of cell cycle and action of tumor suppressor proteins and is also a target of the transforming proteins of small DNA tumor viruses. The E2F proteins contain several evolutionally conserved domains found in most members of the family. These domains include a DNA binding domain, a dimerization domain which determines interaction with the differentiation regulated transcription factor proteins (DP), a transactivation domain enriched in acidic amino acids, and a tumor suppressor protein association domain which is embedded within the transactivation domain. This protein and another 2 members, E2F1 and E2F2, have an additional cyclin binding domain. This protein binds specifically to retinoblastoma protein pRB in a cell-cycle dependent manner. [provided by RefSeq

Sequence: MQSGGGVKTDDTSTLNSLCGYAWVYVWEEKQRCRLSSFFSSSASIPGLLPSHTLDLVQNVGVVLDEALGWGRERELCVKCLLEMHCGVFSCMGNHLCQAFPHFPYLSHLVSCLCFQLCVILFASCTKLIFSKV

Specifications

Antigen E2F3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length recombinant E2F3.
Formulation 50% glycerol
Gene E2F3
Gene Accession No. BC016847
Gene Alias DKFZp686C18211/E2F-3/KIAA0075/MGC104598
Gene Symbols E2F3
Host Species Mouse
Immunogen E2F3 (AAH16847, 1 a.a. to 133 a.a) full-length recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1871
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.