Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-EBI3 (Neutralizing), Clone: V1.4H6.25,

Catalog No. MABF836MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF836MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF836MI Supplier MilliporeSigma™ Supplier No. MABF836
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects EBI3 (Neutralizing) clone: V1.4H6.25 in Mouse samples, and it is validated for Neutralization, Western Blotting

EBI3/IL27B is the beta subunit of the functional cytokine IL-27. The beta subunit is required for activity. IL-27 is a critical cytokine of mammalian innate immune response. IL-27 has both pro and anti-inflammatory activities. IL-27 regulates T-helper cell development and differentiation into TH1, TH2 and TH17 effector cells. IL-27 also is important in isotype switching for B-cells, and it drives rapid clonal expansion of native but not memory CD4 cells. IL-27 is also important for its anti-tumor and anti-angiogenic activity on tumor and cancer cells. The beta subunit EBI3/IL27B is also a component of IL-35 which is a heterodimer of IL-12alpha and IL-27B/EBI3. IL-35 plays a role in immune suppression. IL-27 is a secreted cytokine, thus IL-27beta subunit is expressed intracellularly associated with the endoplasmic reticulum and membrane vesicles and is detected extracellularly as part of the mature cytokine as well.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EBI3 (Neutralizing)
Applications Neutralization, Western Blot
Classification Monoclonal
Clone V1.4H6.25
Formulation Purified mouse monoclonal IgG3κ in buffer containing PBS without preservatives.
Gene Accession No. O35228
Gene Symbols Ebi3; Il27b
Host Species Mouse
Immunogen Recombinant protein corresponding to mouse EBI3/IL-35.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_056581
Target Species Mouse
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Purified
Isotype IgG3 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.